missing translation for 'onlineSavingsMsg'
Learn More

POLR2D Antibody [PE], Novus Biologicals™

Product Code. 30763823 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763823 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763823 Supplier Novus Biologicals Supplier No. NBP347092PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POLR2D Polyclonal antibody specifically detects POLR2D in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2D
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias DNA-directed RNA polymerase II 16 kDa polypeptide, DNA-directed RNA polymerase II subunit D, DNA-directed RNA polymerase II subunit RPB4, HSRBP4, HSRPB4, polymerase (RNA) II (DNA directed) polypeptide D, RBP4, RNA polymerase II 16 kDa subunit, RNA polymerase II subunit B4, RNA polymerase II subunit hsRBP4, RPB16
Host Species Rabbit
Immunogen polymerase(RNA) II(DNA directed) polypeptide D. (Uniprot# O15514)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5433
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.