missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR2C Recombinant Rabbit Monoclonal Antibody (3T8K8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542940
This item is not returnable.
View return policy
Description
Positive test controls include: 293T, Mouse kidney, Rat lung, Rat brain, Rat spleen. Immunogen sequence: MPYANQPTVR ITELTDENVK FIIENTDLAV ANSIRRVFIA EVPIIAIDWV QIDANSSVLH DEFIAHRLGL IPLISDDIVD KLQYSRDCTC EEFCPECSVE.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB3 is part of the core element with the central large cleft and the clamp element that moves to open and close the cleft.
Specifications
| POLR2C | |
| Recombinant Monoclonal | |
| 0.37 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19387, P97760 | |
| POLR2C | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RPB3/POLR2C (P19387). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3T8K8 | |
| Unconjugated | |
| POLR2C | |
| 33kDa; A-152E5.7; DNA-directed RNA polymerase II 33 kDa polypeptide; DNA-directed RNA polymerase II subunit C; DNA-directed RNA polymerase II subunit RPB3; hRPB33; hsRPB3; mRBP31; Polr2c; polymerase (RNA) II (DNA directed) polypeptide C; polymerase (RNA) II (DNA directed) polypeptide C, 33kDa; polymerase (RNA) II subunit C; polymerase II polypeptide C; RNA polymerase II 3; RNA polymerase II subunit 3; RNA polymerase II subunit B3; RNA polymerase II subunit C; RPB3; RPB31; RPB33; Rpo2-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20021, 361365, 5432 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction