Learn More
Invitrogen™ POLR2C Recombinant Rabbit Monoclonal Antibody (24GB1535)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557235
 View more versions of this product
Product Code. 30274844
  • £434.00 / 100µL
Description
POLR2C Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB3 is part of the core element with the central large cleft and the clamp element that moves to open and close the cleft.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POLR2C
Recombinant Monoclonal
Unconjugated
POLR2C
33kDa; A-152E5.7; DNA-directed RNA polymerase II 33 kDa polypeptide; DNA-directed RNA polymerase II subunit C; DNA-directed RNA polymerase II subunit RPB3; hRPB33; hsRPB3; mRBP31; Polr2c; polymerase (RNA) II (DNA directed) polypeptide C; polymerase (RNA) II (DNA directed) polypeptide C, 33kDa; polymerase (RNA) II subunit C; polymerase II polypeptide C; RNA polymerase II 3; RNA polymerase II subunit 3; RNA polymerase II subunit B3; RNA polymerase II subunit C; RPB3; RPB31; RPB33; Rpo2-3
Rabbit
Affinity chromatography
RUO
20021, 361365, 5432
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1535
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P19387, P97760
POLR2C
A synthesized peptide derived from human POLR2C (aa 50-100).
100 μL
Primary
Human, Mouse, Rat