missing translation for 'onlineSavingsMsg'
Learn More

POLR2C Rabbit anti-Human, Mouse, Rat, Clone: 3T8K8, Novus Biologicals™

Product Code. 18337504 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337504 100 μg 100µL
18373046 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18337504 Supplier Novus Biologicals Supplier No. NBP316871100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

POLR2C Monoclonal antibody specifically detects POLR2C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2C
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3T8K8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DNA-directed RNA polymerase II 33 kDa polypeptide, DNA-directed RNA polymerase II subunit C, DNA-directed RNA polymerase II subunit RPB3, hRPB33, hsRPB3, polymerase (RNA) II (DNA directed) polypeptide C (33kD), polymerase (RNA) II (DNA directed) polypeptide C, 33kDa, RNA polymerase II subunit 3, RNA polymerase II subunit B3, RPB3, RPB31, RPB33
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human POLR2C (P19387).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5432
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.