missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLR2C Polyclonal Antibody
GREENER_CHOICE

Product Code. 16518771
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16518771 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16518771 Supplier Invitrogen™ Supplier No. PA595927

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MPYANQPTVR ITELTDENVK FIIENTDLAV ANSIRRVFIA EVPIIAIDWV QIDANSSVLH DEFIAHRLGL IPLISDDIVD KLQYSRDCTC EEFCPECSVE FTLDVRCNED QTRHVTSRDL ISNSPRVIPV TSRNRDNDPN DYVEQDDILI VKLRKGQELR LRAYAKKGFG KEHAKWNPTA GVAFEYDPDN ALRHTVYPKP EEWPKSEYSE LDEDESQAPY DPNGKPERFY YNVESCGSLR PETIVLSALS GLKKKLSDLQ TQLSHEIQSD VLTIN; Positive Samples: SW620, OVCAR3, Mouse spleen; Cellular Location: Nucleus.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB3 is part of the core element with the central large cleft and the clamp element that moves to open and close the cleft.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2C
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLR2C
Gene Accession No. P19387, P97760
Gene Alias 33kDa; A-152E5.7; DNA-directed RNA polymerase II 33 kDa polypeptide; DNA-directed RNA polymerase II subunit C; DNA-directed RNA polymerase II subunit RPB3; hRPB33; hsRPB3; mRBP31; Polr2c; polymerase (RNA) II (DNA directed) polypeptide C; polymerase (RNA) II (DNA directed) polypeptide C, 33kDa; polymerase (RNA) II subunit C; polymerase II polypeptide C; RNA polymerase II 3; RNA polymerase II subunit 3; RNA polymerase II subunit B3; RNA polymerase II subunit C; RPB3; RPB31; RPB33; Rpo2-3
Gene Symbols POLR2C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human RPB3/POLR2C (NP_1165581).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20021, 5432
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.