missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2C Antibody (SR1551), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | POLR2C |
|---|---|
| Clone | SR1551 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653434
|
Novus Biologicals
NBP3-22138-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681465
|
Novus Biologicals
NBP3-22138-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POLR2C Monoclonal antibody specifically detects POLR2C in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| POLR2C | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DNA-directed RNA polymerase II 33 kDa polypeptide, DNA-directed RNA polymerase II subunit C, DNA-directed RNA polymerase II subunit RPB3, hRPB33, hsRPB3, polymerase (RNA) II (DNA directed) polypeptide C (33kD), polymerase (RNA) II (DNA directed) polypeptide C, 33kDa, RNA polymerase II subunit 3, RNA polymerase II subunit B3, RPB3, RPB31, RPB33 | |
| A synthesized peptide derived from human POLR2C (Uniprot #: P19387) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1551 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5432 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title