missing translation for 'onlineSavingsMsg'
Learn More

POLR2C Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £366.00

Specifications

Antigen POLR2C
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:20-1:50
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18691002
View Documents
Novus Biologicals
NBP2-94433-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18689941
View Documents
Novus Biologicals
NBP2-94433-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

POLR2C Polyclonal antibody specifically detects POLR2C in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

POLR2C
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
DNA Repair, DNA replication Transcription Translation and Splicing
PBS (pH 7.3), 50% glycerol
5432
IgG
Affinity purified
Western Blot 1:500-1:2000, Immunoprecipitation 1:20-1:50
Polyclonal
Purified
RUO
Human, Mouse
DNA-directed RNA polymerase II 33 kDa polypeptide, DNA-directed RNA polymerase II subunit C, DNA-directed RNA polymerase II subunit RPB3, hRPB33, hsRPB3, polymerase (RNA) II (DNA directed) polypeptide C (33kD), polymerase (RNA) II (DNA directed) polypeptide C, 33kDa, RNA polymerase II subunit 3, RNA polymerase II subunit B3, RPB3, RPB31, RPB33
Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human POLR2C (NP_116558.1). MPYANQPTVRITELTDENVKFIIENTDLAVANSIRRVFIAEVPIIAIDWVQIDANSSVLHDEFIAHRLGLIPLISDDIVDKLQYSRDCTCEEFCPECSVEFTLDVRCNEDQTRHVTSRDLISNSPRVIPVTSRNRDNDPNDYVEQDDILIVKLRKGQELRLRAYAKKGFGKEHAKWNPTAGVAFEYDPDNALRHTVYPKPEEWPKSEYSELDEDESQAPYDPNGKPERFYYNVESCGSLRPETIVLSALSGLKKKLSDLQTQLSHEIQSDVLTIN
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.