missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20370-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to POLR2B.
This gene encodes the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA.Specifications
| POLR2B | |
| Polyclonal | |
| Unconjugated | |
| POLR2B | |
| hRPB140, hsRPB2, POL2RB, POLR2B, RNA polymerase II subunit 2, RNA polymerase II subunit B2, RPB2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231329, 289561, 5431 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30876, Q8CFI7 | |
| Polr2b | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction