missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557980
This item is not returnable.
View return policy
Description
Immunogen sequence: FFRSVFYRSY KEQESKKGFD QEEVFEKPTR ETCQGMRHAI YDKLDDDGLI APGVRVSGDD VIIGKTVTLP ENEDELESTN RRYTKRDCST FLRTSE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
This gene encodes the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA.
Specifications
| POLR2B | |
| Polyclonal | |
| Unconjugated | |
| POLR2B | |
| DNA directed RNA polymerase II polypeptide B; DNA-directed RNA polymerase II 140 kDa polypeptide; DNA-directed RNA polymerase II subunit B; DNA-directed RNA polymerase II subunit RPB2; hRPB140; hs hRPB140; hsRPB2; Pol2rb; POLR2B; polymerase (RNA) II (DNA directed) polypeptide B; polymerase (RNA) II (DNA directed) polypeptide B, 140kDa; polymerase (RNA) II subunit B; RNA polymerase II second largest subunit; RNA polymerase II subunit 2; RNA polymerase II subunit B; RNA polymerase II subunit B2; rpb 2; Rpb140; Rpb2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P30876 | |
| POLR2B | |
| Recombinant protein corresponding to Human POLR2B. Recombinant protein control fragment (Product #RP-96595). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction