missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR1E Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530802
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HepG2, HCT116. Predicted reactivity: Chimpanzee (100%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors (PubMed:24207024). Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF (PubMed:24207024). [UniProt].
Specifications
| POLR1E | |
| Polyclonal | |
| Unconjugated | |
| Polr1e | |
| 53kDa; AU042259; D030019D19Rik; DNA-directed RNA polymerase I subunit E; DNA-directed RNA polymerase I subunit RPA49; FLJ13390; FLJ13970; hypothetical protein LOC511587; Paf53; Polr1e; polymerase (RNA) I associated factor 1; polymerase (RNA) I polypeptide E; polymerase (RNA) I polypeptide E, 53kDa; polymerase (RNA) I subunit E; PRAF1; RGD1565773; RNA polymerase I associated factor (Paf53); RNA polymerase I associated factor 53; RNA polymerase I subunit A49; RNA polymerase I subunit E; RNA polymerase I-associated factor 1; RNA polymerase I-associated factor 53; RP11-405L18.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64425 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9GZS1 | |
| Polr1e | |
| Recombinant protein encompassing a sequence within the center region of human PRAF1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction