missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLR1C Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA587402

Product Code. 16505901

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAASQAVEEM RSRVVLGEFG VRNVHTTDFP GNYSGYDDAW DQDRFEKNFR VDVVHMDENS LEFDMVGIDA AIANAFRRIL LAEVPTMAVE KVLVYNNTSI VQDEILAHRL GLIPIHADPR LFEYRNQGDE EGTEIDTLQF RLQVRCTRNP HAAKDSSDPN ELYVNHKVYT RHMTWIPLGN QADLFPEGTI RPVHDDILIA; Positive Samples: K-562, 293T, MCF7; Cellular Location: Nucleus.

RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA16, RPA40 and RPA135 are subunits of Pol I that associate with each other at an early stage of RNA Pol I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POLR1C
Polyclonal
Unconjugated
POLR1C
40kDa; AA409007; AA959927; Ac2-127; AC40; AL024089; DNA-directed RNA polymerase I subunit C; DNA-directed RNA polymerases I and III 40 kDa polypeptide; DNA-directed RNA polymerases I and III subuni RPAC1-like protein; DNA-directed RNA polymerases I and III subunit RPAC1; HLD11; Polr1c; POLR1E; polymerase (RNA) I polypeptide C; polymerase (RNA) I polypeptide C, 30kDa; polymerase (RNA) I subunit C; RNA polymerase 1-1; RNA polymerase 1-1 (40 kDa subunit); RNA polymerase I 140 kDa subunit; RNA polymerase I and III subunit C; RNA polymerase I subunit; RNA polymerase I subunit C; RNA polymerase subunit I; RNA polymerases I and III subunit AC1; RPA39; RPA40; RPA5; RPAC1; RPC40; Rpo1-1; TCS3; Unknown (protein for MGC:134521); zgc:65973
Rabbit
Affinity chromatography
RUO
9533
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Western Blot
0.98 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15160
POLR1C
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human POLR1C (NP_9760351).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.