missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548383
This item is not returnable.
View return policy
Description
Predicted to react with rat based on sequence homology.
Eukaryotic RNA polymerase I (pol I) is responsible for the transcription of ribosomal RNA (rRNA) genes and production of rRNA, the primary component of ribosomes. Pol I is a multisubunit enzyme composed of 6 to 14 polypeptides, depending on the species. Most of the mass of the pol I complex derives from the 2 largest subunits, Rpa1 and Rpa2 in yeast. POLR1B is homologous to Rpa2 (Seither and Grummt, 1996).
Specifications
| POLR1B | |
| Polyclonal | |
| Unconjugated | |
| Polr1b | |
| 128kDa; A127; D630020H17Rik; DNA-directed RNA polymerase I 135 kDa polypeptide; DNA-directed RNA polymerase I 135kDa polypeptide; DNA-directed RNA polymerase I subunit RPA2; Polr1b; polymerase (RNA) I polypeptide B; polymerase (RNA) I polypeptide B, 128kDa; polymerase (RNA) I subunit B; RNA polymerase 1-2; RNA polymerase 1-2, 128 kDa subunit; RNA polymerase I (127 kDa subunit); RNA polymerase I polypeptide B; RNA polymerase I subunit 2; RNA polymerase I subunit B; RPA116; RPA135; Rpa2; Rpo1-2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 84172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H9Y6 | |
| Polr1b | |
| KLH conjugated synthetic peptide between 89-117 amino acids from the N-terminal region of human POLR1B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction