missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537156
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 195 kDa. Purity is >95% by SDS-PAGE.
SLP-76 was originally identified as a substrate of the ZAP-70 protein tyrosine kinase following T cell receptor (TCR) ligation in the leukemic T cell line Jurkat. The SLP-76 locus has been localized to human chromosome 5q33 and the gene structure has been partially characterized in mice. The human and murine cDNAs both encode 533 amino acid proteins that are 72% identical and comprised of three modular domains. The NH2-terminus contains an acidic region that includes a PEST domain and several tyrosine residues which are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a COOH-terminal SH2 domain. A number of additional proteins have been identified that associate with SLP-76 both constitutively and inducibly following receptor ligation, supporting the notion that SLP-76 functions as an adaptor or scaffold protein. Studies using SLP-76 deficient T cell lines or mice have provided strong evidence that SLP-76 plays a positive role in promoting T cell development and activation as well as mast cell and platelet function.
Specifications
| POLR1A | |
| Polyclonal | |
| Unconjugated | |
| POLR1A | |
| 194kDa; 3010014K16Rik; A190; A194; AFDCIN; DNA-directed RNA polymerase I A; DNA-directed RNA polymerase I largest subunit; DNA-directed RNA polymerase I subunit A; DNA-directed RNA polymerase I subunit A1; DNA-directed RNA polymerase I subunit RPA1; mRPA1; Polr1a; polymerase (RNA) I polypeptide A; polymerase (RNA) I polypeptide A, 194kDa; polymerase (RNA) I subunit A; RNA polymerase 1-4; RNA polymerase 1-4 (194 kDa subunit); RNA polymerase I (194 kDa subunit); RNA polymerase I 194 kDa subunit; RNA polymerase I subunit A; RNA polymerase I subunit A1; Rpa1; RPA194; RPO14; RPO1-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20019, 25885, 83581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35134, O54889, O95602 | |
| POLR1A | |
| A synthetic peptide corresponding to residues in human POLR1A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction