missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ POLR1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29980
This item is not returnable.
View return policy
Description
POLR1A Polyclonal antibody specifically detects POLR1A in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| POLR1A | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25885 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DNA-directed RNA polymerase I subunit RPA1, A190, polymerase (RNA) I polypeptide A, 194kDa, RPA1, RPA194, RPO14, RPO1-4 | |
| Between 1425 and 1475 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction