missing translation for 'onlineSavingsMsg'
Learn More

Poloxin, MedChemExpress

Product Code. 16461237
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461237 50 mg 50mg
16424998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461237 Supplier Medchem Express Supplier No. HY12134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Poloxin is a non-ATP competitive Polo-like Kinase 1 (PLK1) inhibitor that targets the polo-box domain, with an IC50 of appr 4.8 μM.

Poloxin is a non-ATP competitive Polo-like Kinase 1 (PLK1) inhibitor that targets the polo-box domain, with an IC50 of appr 4.8 μM.

Specifications

Chemical Name or Material Poloxin
Physical Form Solid
CAS 321688-88-4
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H19NO3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 14.29 mg/mL (48.06 mM; Need ultrasonic)
SMILES O=C(C(C(C)C)=C/1)C=C(C)C1=N/OC(C2=CC=CC=C2C)=O
Molecular Weight (g/mol) 297.35
Formula Weight 297.35
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.