missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566816
This item is not returnable.
View return policy
Description
Immunogen sequence: SEATHVVMEET SAEEAVSWQE RRMAAAPPGC TPPALLDISW LTESLGAGQP VPVECRHRLE VAGPRKGPLS PAWMPAYACQ RPTPLTHHNT G Highest antigen sequence identity to the following orthologs - mouse 70%, rat 68%.
Gap-filling polymerase involved in repair of DNA double-strand breaks by non-homologous end joining (NHEJ). Participates in immunoglobulin (Ig) light chain gene rearrangement in V(D)J recombination.
Specifications
| POLM | |
| Polyclonal | |
| Unconjugated | |
| POLM | |
| AI462166; B230309I03Rik; DNA polymerase mu; DNA-directed DNA polymerase mu; DNA-directed DNA/RNA polymerase mu; Pol iota; pol Mu; Polm; polmu; polymerase (DNA directed), mu; polymerase (DNA) mu; polymerase (DNA-directed), mu; Tdt-N; Terminal transferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27434 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NP87 | |
| POLM | |
| Recombinant Human POLM. Recombinant protein control fragment (Product #RP-105377). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction