missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599800
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total POLM.
Gap-filling polymerase involved in repair of DNA double-strand breaks by non-homologous end joining (NHEJ). Participates in immunoglobulin (Ig) light chain gene rearrangement in V(D)J recombination.
Specifications
| POLM | |
| Polyclonal | |
| Unconjugated | |
| POLM | |
| AI462166; B230309I03Rik; DNA polymerase mu; DNA-directed DNA polymerase mu; DNA-directed DNA/RNA polymerase mu; Pol iota; pol Mu; Polm; polmu; polymerase (DNA directed), mu; polymerase (DNA) mu; polymerase (DNA-directed), mu; Tdt-N; Terminal transferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27434 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NP87 | |
| POLM | |
| A synthesized peptide derived from human POLM(Accession Q9NP87), corresponding to amino acid residues L298-P348. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction