missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLM Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566220
This item is not returnable.
View return policy
Description
Immunogen sequence: ITHPKEGQEAG LLPRVMCRLQ DQGLILYHQH QHSCCESPTR LAQQSHMDAF ERSFCIFRLP QPPGAAVGGS TRPCPSWKAV RVDLVVAPVS QFP Highest antigen sequence identity to the following orthologs - mouse 66%, rat 67%.
Gap-filling polymerase involved in repair of DNA double-strand breaks by non-homologous end joining (NHEJ). Participates in immunoglobulin (Ig) light chain gene rearrangement in V(D)J recombination.
Specifications
| POLM | |
| Polyclonal | |
| Unconjugated | |
| POLM | |
| AI462166; B230309I03Rik; DNA polymerase mu; DNA-directed DNA polymerase mu; DNA-directed DNA/RNA polymerase mu; Pol iota; pol Mu; Polm; polmu; polymerase (DNA directed), mu; polymerase (DNA) mu; polymerase (DNA-directed), mu; Tdt-N; Terminal transferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27434 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NP87 | |
| POLM | |
| Recombinant Human POLM. Recombinant protein control fragment (Product #RP-106901). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto