missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pollen allergen Phl p 1 Human anti-Plant, Clone: 25, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Pollen allergen Phl p 1 Monoclonal specifically detects Pollen allergen Phl p 1 in Plant samples. It is validated for Western Blot, ELISA, Dot Blot.
Specifications
Specifications
| Antigen | Pollen allergen Phl p 1 |
| Applications | Western Blot, ELISA, Dot Blot |
| Classification | Monoclonal |
| Clone | 25 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Dot Blot |
| Formulation | PBS |
| Gene Accession No. | P43213 |
| Gene Alias | Allergen Phl p I, grass pollen allergen |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?