missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113372
This item is not returnable.
View return policy
Description
POLL Polyclonal Antibody for Western Blot, ICC/IF, ELISA
The protein encoded by this gene functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation. Alternatively spliced transcript variants have been found for this gene.
Specifications
| POLL | |
| Polyclonal | |
| Unconjugated | |
| Poll | |
| 1110003P06Rik; AV007317; BETAN; DNA polymerase beta-2; DNA polymerase beta-N; DNA polymerase kappa; DNA polymerase lambda; DNA-directed DNA polymerase lambda; EC 2.7.7.7; FLJ46002; OTTHUMP00000020323; Pol beta2; Pol Lambda; Polk; POLKAPPA; POL-KAPPA; Poll; polymerase (DNA directed), lambda; polymerase (DNA) lambda; RP11-529I10.1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 27343, 361767 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5RKI3, Q9UGP5 | |
| Poll | |
| Recombinant Human DNA polymerase lambda protein (16-211AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction