missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLK Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543223
This item is not returnable.
View return policy
Description
Peptide sequence: KINKIIMEAT KGSRFYGNEL KKEKQVNQRI ENMMQQKAQI TSQQLRKAQL Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
External and internal DNA-damaging agents continually threaten the integrity of genetic material in cells. Although a variety of repair mechanisms exist to remove the resulting lesions, some lesions escape repair and block the replication machinery. Members of the Y family of DNA polymerases, such as POLK, permit the continuity of the replication fork by allowing replication through such DNA lesions. Each Y family polymerase has a unique DNA-damage bypass and fidelity profile. POLK is specialized for the extension step of lesion bypass.
Specifications
| POLK | |
| Polyclonal | |
| Unconjugated | |
| POLK | |
| DinB homolog 1; DINB protein; Dinb1; DINP; DNA damage-inducible protein b; DNA polymerase kappa; DNA-directed DNA polymerase kappa; LOW QUALITY PROTEIN: DNA polymerase kappa; POLK; POLQ; polymerase (DNA directed) kappa; polymerase (DNA directed), kappa; polymerase (DNA) kappa | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51426 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UBT6 | |
| POLK | |
| Synthetic peptide directed towards the N-terminal of human POLK (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction