missing translation for 'onlineSavingsMsg'
Learn More

Poliovirus 3 Antibody (45 D5), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18426097 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426097 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426097 Supplier Novus Biologicals Supplier No. NBP105102JF549

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Poliovirus 3 Monoclonal antibody specifically detects Poliovirus 3 in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Poliovirus 3
Applications ELISA
Classification Monoclonal
Clone 45 D5
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias PV-3
Host Species Mouse
Immunogen Poliovirus type 3 (inactivated)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.