missing translation for 'onlineSavingsMsg'
Learn More

Polidocanol, MedChemExpress

Product Code. 16462369
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462369 1 g 1g
16441148 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462369 Supplier Medchem Express Supplier No. HYB2106
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Polidocanol is a sclerosing agent used successfully to treat extremity and esophageal varices and telangiectasias.

Polidocanol is a sclerosing agent used successfully to treat extremity and esophageal varices and telangiectasias.

Specifications

Chemical Name or Material Polidocanol
CAS 9002-92-0
Color White
Purity Grade Notes Research
Molecular Formula (C2H4O)nC12H26O
Quantity 1 g
Synonym Polyoxyethylene lauryl ether Polyoxyethyleneglycol Dodecyl Ether
Solubility Information H2O : 50 mg/mL (Need ultrasonic) ∣DMSO : 50 mg/mL (Need ultrasonic)
SMILES OCCCOCCCCCCCCCCCCC.[n]
Percent Purity 97.0%
Grade Research
Physical Form Crystals
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.