missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLI Recombinant Rabbit Monoclonal Antibody (HL3115)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556680
This item is not returnable.
View return policy
Description
POLI Recombinant Monoclonal Antibody for Western Blot
Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunoglobulin genes. Forms a Schiff base with 5'-deoxyribose phosphate at abasic sites, but may not have lyase activity.
Specifications
| POLI | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q6R3M4, Q9UNA4 | |
| Poli | |
| Recombinant fragment of human DNA polymerase iota. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3115 | |
| Unconjugated | |
| Poli | |
| DNA polymerase iota; eta2; Poli; polymerase (DNA directed) iota; polymerase (DNA directed), iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B; RAD30B; RAD3OB | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11201, 26447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction