Learn More
Invitrogen™ POLI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595515
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human placenta tissue, human A549 whole cell, human SK-OV-3 whole cell, rat testis tissue, rat testis tissue, rat kidney tissue, rat stomach tissue, mouse testis tissue, mouse testis tissue, mouse kidney tissue, mouse stomach tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunoglobulin genes. Forms a Schiff base with 5'-deoxyribose phosphate at abasic sites, but may not have lyase activity.
Specifications
| POLI | |
| Polyclonal | |
| Unconjugated | |
| Poli | |
| DNA polymerase iota; eta2; Poli; polymerase (DNA directed) iota; polymerase (DNA directed), iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B; RAD30B; RAD3OB | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11201, 26447, 291526 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q6R3M4, Q9UNA4 | |
| Poli | |
| A synthetic peptide corresponding to a sequence of human DNA Polymerase iota (DIDPQVFYELPEAVQKELLAEWKRAGSDFHIGHK). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.