missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLI Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA316539
This item is not returnable.
View return policy
Description
Suggested positive control: HeLa whole cell extract (100 μg) or GM0193 (lymphoblastoid cell line)..
Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunoglobulin genes. Forms a Schiff base with 5'-deoxyribose phosphate at abasic sites, but may not have lyase activity.
Specifications
| POLI | |
| Polyclonal | |
| Unconjugated | |
| Poli | |
| DNA polymerase iota; eta2; Poli; polymerase (DNA directed) iota; polymerase (DNA directed), iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B; RAD30B; RAD3OB | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.02% sodium azide | |
| Q6R3M4, Q9UNA4 | |
| Poli | |
| A 15-mer peptide in the C-terminal region of human DNA Polymerase Iota, conjugated to KLH. | |
| RUO | |
| 11201, 26447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction