missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552332
This item is not returnable.
View return policy
Description
Immunogen sequence: PPLTMLFLCA TKFSASAPSS STDITSFLSS DPSSLPKVPV TSSEAKTQGS GPAVTATKKA TTSLESFFQK AAERQKVKEA SLSSLTAPTQ APMSNSPSKP SLPFQTSQST GTE Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 61%.
The XPA gene encodes a zinc metalloprotein that preferentially binds to DNA damaged by UV radiation and chemical carcinogens and is required for the incision step during nucleotide excision repair The XPB and XPD genes encode DNA helicases involved in several DNA metabolic pathways, including DNA repair and transcription, and the XPG gene product is an endonuclease that cuts on the 3' side of a DNA lesion during nucleotide excision repair. Molecular defects in the XP variant (POL H) group maintain normal excision repair, yet they result in a substantial reduction in the ability to synthesize intact daughter DNA strands during DNA replication following DNA damage.
Specifications
| POLH | |
| Polyclonal | |
| Unconjugated | |
| POLH | |
| DNA polymerase eta; DNA-directed DNA polymerase eta; FLJ16395; FLJ21978; Polh; polymerase (DNA directed), eta; polymerase (DNA directed), eta (RAD 30 related); polymerase (DNA) eta; RAD30; RAD30 homolog A; RAD30A; Xeroderma pigmentosum variant type protein; xeroderma pigmentosum variant type protein homolog; XP V; XPV; XP-V | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5429 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y253 | |
| POLH | |
| Recombinant protein corresponding to Human POLH. Recombinant protein control fragment (Product #RP-89494). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction