missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10997-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the processivity subunit of the mitochondrial DNA polymerase gamma. The encoded protein forms a heterotrimer containing one catalytic subunit and two processivity subunits. This protein enhances DNA binding and promotes processive DNA synthesis. Mutations in this gene result in autosomal domit progressive external ophthalmoplegia with mitochondrial DNA deletions.Specifications
| POLG2 | |
| Polyclonal | |
| Unconjugated | |
| POLG2 | |
| HP55, MTPOLB, p55, PEOA4, POLB, POLG BETA, POLG2, POLGB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11232, 303612, 50776 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9QZM2, Q9UHN1 | |
| Polg2 | |
| POLG2 Fusion Protein Ag1424 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction