missing translation for 'onlineSavingsMsg'
Learn More

POLG2 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18355433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355433 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355433 Supplier Novus Biologicals Supplier No. NBP299015R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POLG2 Polyclonal specifically detects POLG2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLG2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DNA polymerase gamma accessory 55 kDa subunit, DNA polymerase subunit gamma-2, mitochondrial, EC 2.7.7.7, HP55, Mitochondrial DNA polymerase accessory subunit, mitochondrial DNA polymerase subunit gamma-2, mitochondrial DNA polymerase, accessory subunit, MtPolB, MTPOLBPOLG-BETA, p55, PEOA4, POLB, POLGB, PolG-beta, polymerase (DNA directed), gamma 2, accessory subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human POLG2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11232
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.