missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLG Recombinant Rabbit Monoclonal Antibody (24GB1085)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557386
This item is not returnable.
View return policy
Description
POLG Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Mitochondrial DNA polymerase is heterotrimeric, consisting of a homodimer of accessory subunits plus a catalytic subunit. The protein encoded by this gene is the catalytic subunit of mitochondrial DNA polymerase. The encoded protein contains a polyglutamine tract near its N-terminus that may be polymorphic. Defects in this gene are a cause of progressive external ophthalmoplegia with mitochondrial DNA deletions 1 (PEOA1), sensory ataxic neuropathy dysarthria and ophthalmoparesis (SANDO), Alpers-Huttenlocher syndrome (AHS), and mitochondrial neurogastrointestinal encephalopathy syndrome (MNGIE). Two transcript variants encoding the same protein have been found for this gene.
Specifications
| POLG | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54098, P54099, Q9QYV8 | |
| POLG | |
| A synthesized peptide derived from human DNA Polymerase gamma (aa 1190-1220). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1085 | |
| Unconjugated | |
| POLG | |
| AA409516; DNA polymerase gamma; DNA polymerase gamma subunit 1 (Mitochondrial DNA polymerase catalytic subunit) (PolG-alpha); DNA polymerase gamma, catalytic subunit; DNA polymerase subunit gamma-1; DNA-directed DNA polymerase gamma; FLJ27114; MDP1; Mip1; MIRAS; Mitochondrial DNA polymerase catalytic subunit; mitochondrial DNA polymerase gamma; mitochondrial DNA polymerase gamma catalytic subunit; mitochondrial DNA polymerase-gamma; mitochondrial polymerase gamma; MTDPS4A; MTDPS4B; PEO; Pol gamma; Polg; POLG1; POLGA; PolG-alpha; polymerase (DNA directed), gamma; polymerase (DNA) gamma, catalytic subunit; polymerase, gamma; SANDO; SCAE; truncated mitochondrial DNA polymerase gamma catalytic subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18975, 5428, 85472 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction