missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584367
This item is not returnable.
View return policy
Description
Immunogen sequence: HLQRHNHLLW LSPTARPDLG GKEADDNCLV MEFDDQATVE INSSGCYSTV CVELDLQNLA VNTILQSHHV NDMEGADSMG ISFDVIQ Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.
POLE (Polepsi) is present in proliferating cells and is implicated in repair of UV-damaged DNA.
Specifications
| POLE | |
| Polyclonal | |
| Unconjugated | |
| POLE | |
| CRCS12; DNA polymerase epsilon catalytic subunit A; DNA polymerase epsilon catalytic subunit protein; DNA polymerase epsilon, catalytic subunit; DNA polymerase II subunit A; DNA-directed DNA polymerase epsilon; FILS; Pole; POLE1; pol-epsilon; polymerase (DNA directed), epsilon; polymerase (DNA directed), epsilon, catalytic subunit; polymerase (DNA) epsilon, catalytic subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5426 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q07864 | |
| POLE | |
| Recombinant protein corresponding to Human POLE. Recombinant protein control fragment (Product #RP-106829). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction