missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578113
This item is not returnable.
View return policy
Description
Positive Control: K562 Predicted Reactivity: Mouse (90%), Rat (90%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
POLE (Polepsi) is present in proliferating cells and is implicated in repair of UV-damaged DNA.
Specifications
| POLE | |
| Polyclonal | |
| Unconjugated | |
| POLE | |
| CRCS12; DNA polymerase epsilon catalytic subunit A; DNA polymerase epsilon catalytic subunit protein; DNA polymerase epsilon, catalytic subunit; DNA polymerase II subunit A; DNA-directed DNA polymerase epsilon; FILS; Pole; POLE1; pol-epsilon; polymerase (DNA directed), epsilon; polymerase (DNA directed), epsilon, catalytic subunit; polymerase (DNA) epsilon, catalytic subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5426 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.45 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q07864 | |
| POLE | |
| Recombinant protein encompassing a sequence within the N-terminus region of human POLE. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction