missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLDIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583925
This item is not returnable.
View return policy
Description
Immunogen sequence: YKKYNNRCLD GQPMKCNLHM NGNVITSDQP ILLRLSDSPS MKKESELPRR VNSASSSNPP AEVDPDTILK ALFKSSGASV TTQPTEFK Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 93%.
This gene encodes a protein that interacts with the DNA polymerase delta p50 subunit. This protein is a specific target of S6 kinase 1 and regulates cell growth. Two transcript variants that encode different protein isoforms have been identified.
Specifications
| POLDIP3 | |
| Polyclonal | |
| Unconjugated | |
| POLDIP3 | |
| 1110008P04Rik; 46 kDa DNA polymerase delta interaction protein; AA408269; AL022852; C77954; DNA polymerase delta interacting protein 3; KIAA1649; mKIAA1649; p46; PDIP46; POLDIP3; polymerase (DNA) delta interacting protein 3; polymerase (DNA-directed), delta interacting protein 3; polymerase delta interacting protein 46; polymerase delta-interacting protein 3; RNA-binding protein P46; S6K1 Aly/REF-like target; SKAR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84271 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BY77 | |
| POLDIP3 | |
| Recombinant protein corresponding to Human POLDIP3. Recombinant protein control fragment (Product #RP-106204). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction