missing translation for 'onlineSavingsMsg'
Learn More

Poldine Metilsulfate, LoGiCal

Product Code. 30458518
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30458518

Brand: LoGiCal LGCFOR1911.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Poldine Metilsulfate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Poldine Metilsulfate
CAS 545-80-2
Analyte or Component Names Poldine Metilsulfate
Shipping Condition Room Temperature
Molecular Formula C21 H26 N O3 . C H3 O4 S
Certifications/Compliance ISO 9001
Synonym Pyrrolidinium, 2-[[(2-hydroxy-2,2-diphenylacetyl)oxy]methyl]-1,1-dimethyl-, methyl sulfate (1:1), 2-(Hydroxymethyl)-1,1-dimethylpyrrolidinium methyl sulfate, benzilate (6 CI,7 CI), Pyrrolidinium, 2-(hydroxymethyl)-1,1-dimethyl-, methyl sulfate, benzilate (8 CI), Pyrrolidinium, 2-[[(hydroxydiphenylacetyl)oxy]methyl]-1,1-dimethyl-, methyl sulfate (salt) (9 CI), (1-Methyl-2-pyrrolidinyl)methyl benzilate methyl methosulfate, Alfaland, CLB 499, IS 499, McN-R 726-47, Nactate, Nacton, Poldine methosulfate, Poldine methyl methosulfate, Poldine methylsulfate, Poldine metilsulfate
InChI Formula InChI=1 S/C21H26NO3.CH4O4S/c1-22(2)15-9-14-19(22)16-25-20(23)21(24,17-10-5-3-6-11-17)18-12-7-4-8-13-18;1-5-6(2,3)4/h3-8,10-13,19,24 H,9,14-16H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1
SMILES COS(=O)(=O)[O-].C[N+]1(C)CCCC1COC(=O)C(O)(c2ccccc2)c3ccccc3
IUPAC Name (1,1-dimethylpyrrolidin-1-ium-2-yl)methyl 2-hydroxy-2,2-diphenylacetate;methyl sulfate
Industry Type Forensics
Molecular Weight (g/mol) 451.53
Quantity 5 mg
Formula Weight 451.1665 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.