missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLD3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596616

Product Code. 16569451

  • £447.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ASKAAAKTQE TNKETKTEAK EVTNASAAGN KAPGKGNMMS NFFGKAAMNK FKVNLDSEQA VKEEKIVEQP TVSVTEPKLA TPAGLKKSSK KAEPVKVLQK EKKRGKRVAL SDDETKETEN MRKKRRRIKL PESDSSEDEV FPDSPGAYEA ESPSPPPPPS PPLEPVPKTE PEPPSVKSSS GENKRKRKRV LKSKTYLDGE GCIVTEKVYE SESCTDSEEE LNMKTSSVHR PPAMTVKKEP REERKGPKKG TAALGKANRQ VSITGFFQRK; Positive Samples: THP-1, HepG2, HeLa, MCF7, H460, Mouse brain; Cellular Location: Nucleus.

DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases. In DNA replication, exonucleases are involved in the processing of Okazaki fragments, whereas in DNA repair, they function to excise damaged DNA fragments and correct recombinational mismatches. These exonucleases include the family of DNA polymerases, namely DNA pol alpha, beta, delta and epsilon. DNA pol delta and DNA pol epsilon are multisubunit enzymes, with DNA pol delta consisting of two subunits which interact with the sliding DNA clamp protein PCNA. DNA pol delta 3, also known as POLD3 or p66, is a 466 amino acid nuclear protein that is required for the proper function of DNA pol delta.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POLD3
Polyclonal
Unconjugated
POLD3
2410142G14Rik; C85233; DNA pol delta3; DNA polymerase delta 3, accessory subunit; DNA polymerase delta subunit 3; DNA polymerase delta subunit C; DNA polymerase delta subunit p66; DNA polymerase delta subunit p68; DNA-directed DNA polymerase delta 3; DNApol delta3; GC12; KIAA0039; P66; P68; Pol delta C subunit (p66); POLD3; polymerase (DNA directed), delta 3; polymerase (DNA) delta 3, accessory subunit; polymerase (DNA-directed), delta 3, accessory subunit; PPP1R128; protein phosphatase 1, regulatory subunit 128
Rabbit
Affinity Chromatography
RUO
10714, 67967
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15054, Q9EQ28
POLD3
Recombinant fusion protein containing a sequence corresponding to amino acids 197-466 of human POLD3 (NP_0065821).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.