missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLD3 Polyclonal Antibody

POLD3 Polyclonal Antibody
Brand: Invitrogen PA536951
This item is not returnable.
View return policy
Description
POLD3 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| POLD3 | |
| Polyclonal | |
| Unconjugated | |
| POLD3 | |
| 2410142G14Rik; C85233; DNA pol delta3; DNA polymerase delta 3, accessory subunit; DNA polymerase delta subunit 3; DNA polymerase delta subunit C; DNA polymerase delta subunit p66; DNA polymerase delta subunit p68; DNA-directed DNA polymerase delta 3; DNApol delta3; GC12; KIAA0039; P66; P68; Pol delta C subunit (p66); POLD3; polymerase (DNA directed), delta 3; polymerase (DNA) delta 3, accessory subunit; polymerase (DNA-directed), delta 3, accessory subunit; PPP1R128; protein phosphatase 1, regulatory subunit 128 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10714, 293144, 67967 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15054, Q9EQ28 | |
| POLD3 | |
| Synthetic peptide corresponding to the 421-466 of human DNA pol delta 3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction