missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLD1 Recombinant Rabbit Monoclonal Antibody (K01_1K93)

Product Code. 30829665
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829665 50 μL
30829212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829665 Supplier Invitrogen™ Supplier No. MA564597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

POLD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

This gene encodes the 125-kDa catalytic subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3' to 5' exonuclease activity and plays a critical role in DNA replication and repair. Alternatively spliced transcript variants have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLD1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1K93
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene POLD1
Gene Accession No. O54747, P28340, P52431
Gene Alias 125kDa; CDC2; CDC2 homolog; CRCS10; DAN polymerase delta 1, catalytic subunit; DNA polymerase delta 1, catalytic domain; DNA polymerase delta catalytic subunit; DNA polymerase delta, catalytic subunit; DNA polymerase subunit delta p125; DNA-directed DNA polymerase delta 1; MDPL; POL D; POLD; POLD1; polymerase (DNA directed), delta 1, catalytic subunit; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit; Polymerase D
Gene Symbols POLD1
Host Species Rabbit
Immunogen A synthetic peptide of human DNA Polymerase delta, catalytic subunit.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18971, 5424, 59294
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.