missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15646-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the 125-kDa catalytic subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3′ to 5′ exonuclease activity and plays a critical role in DNA replication and repair. Alternatively spliced transcript variants have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.Specifications
| POLD1 | |
| Polyclonal | |
| Unconjugated | |
| POLD1 | |
| CDC2, POLD, POLD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18971, 5424, 59294 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54747, P28340, P52431 | |
| Pold1 | |
| POLD1 Fusion Protein Ag8178 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction