missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLD1 Monoclonal Antibody (607)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA123200
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, HeLa, BCL-1.
This gene encodes the 125-kDa catalytic subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3' to 5' exonuclease activity and plays a critical role in DNA replication and repair. Alternatively spliced transcript variants have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.
Specifications
| POLD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 20% glycerol and no preservative; pH 7 | |
| P28340, P52431 | |
| POLD1 | |
| Recombinant protein encoding amino acids 1-140 of the large subunit of human DNA polymerase d. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 607 | |
| Unconjugated | |
| POLD1 | |
| 125kDa; CDC2; CDC2 homolog; CRCS10; DAN polymerase delta 1, catalytic subunit; DNA polymerase delta 1, catalytic domain; DNA polymerase delta catalytic subunit; DNA polymerase delta, catalytic subunit; DNA polymerase subunit delta p125; DNA-directed DNA polymerase delta 1; MDPL; POL D; POLD; POLD1; polymerase (DNA directed), delta 1, catalytic subunit; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit; Polymerase D | |
| Mouse | |
| Protein G | |
| RUO | |
| 18971, 5424 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction