missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Polatuzumab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Tekniske data

Antigen Polatuzumab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Se flere specifikationer

Produkter 2
Produktkode Brand Quantity Pris Mængde & tilgængelighed  
Produktkode Brand Quantity Pris Mængde & tilgængelighed  
30282483
GREENER_CHOICE
missing translation for 'viewDocuments'
Invitrogen™
MA558511
100 μg
£397.00
100µg
Se alternative produkter
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
30284592
missing translation for 'viewDocuments'
Invitrogen™
MA558512
1 mg
£1002.00
1mg
Se alternative produkter
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Beskrivelse

Beskrivelse

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Polatuzumab is a biosimilar that targets CD79B. CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
Tekniske data

Tekniske data

Polatuzumab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Polatuzumab vedotin; polatuzumab vedotin-piiq; POLIVY
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.