missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PolarScreen™ Progesterone Receptor Competitor Assay Kit, Green

Product Code. 15370186
Change view
Click to view available options
Quantity:
400 x 20 μL assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15370186 400 x 20 μL assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15370186 Supplier Invitrogen™ Supplier No. A15905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PolarScreen™ Progesterone Receptor Competitor Assay Kit, Green

The PolarScreen™ Progesterone Receptor Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential progesterone receptor (PR) ligands. The kit uses insect cell-expressed, human PR ligand binding domain tagged with glutathione-S-transferase (GST) (also available separately) and a novel, high-affinity, fluorescent progesterone receptor ligand (Fluormone™ PL Green) in a homogenous, mix-and-read assay format.

This kit produces a green color read-out; a 'red' kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance contact us at drugdiscoverytech@lifetech.com.

The PolarScreen Progesterone Receptor Competitor Assay is:

  • Complete & ready-to-use out of the box—all reagents included, just add your test compounds
  • High throughput compatible—fluorescence polarization assays in a 384-well plate
  • Convenient—components available in bulk sizes for larger screening needs

    How the Assay Works
    An N-terminal fusion of glutathione transferase to the ligand-binding domain of the human progesterone receptor (PR-LBD(GST)) is added to a fluorescently-tagged progesterone ligand (Fluormone PL Green) in the presence of competitor test compounds in microwell plates. The presence of effective competitors prevents the formation of a PL Green/PR-LBD(GST) complex, resulting in a decrease of the polarization value. The shift in polarization value in the presence of test compounds is used to determine relative affinity of test compounds for PR-LBD(GST).

    Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Quantity 400 x 20 μL assays
    Content And Storage 1 tube Progesterone Receptor Ligand Binding Domain Recombinant Protein (store at -68 to -85°C)
    1 tube of Fluormone PL Green (store at -5 to -30°C)
    2 bottles of PR Screening Buffer (store at -5 to -30°C)
    1 tube of 1 M DTT (store at -5 to -30°C)
    Product Line PolarScreen
    For Use With (Application) Competitive Binding Assay, Nuclear Receptor Assay
    Ligand PR
    Product Type Assay Kit
    No. of Assays 400 x 20 μL assays

    For Research Use Only. Not for use in diagnostic procedures.

    Nom du produit
    Sélectionnez un problème

    En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.