missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PolarScreen™ PPARγ-Competitor Assay Kit, Green
Description
The PPARγ (Peroxisome Proliferator-Activated Receptor-gamma) Competitor Assay, Green uses the human-derived recombinant PPARγ ligand-binding domain (PPARγ-LBD) tagged with an N-terminal GST-tag and a novel, tight-binding, selective fluorescent PPARγ ligand (Fluormone™ PPARγ Green) in a homogenous, mix-and-read assay format. PPARγ-LBD/Fluormone™ PPARγ Green complex produces high polarization. This complex is then added to individual test compounds in microwell plates. Competitors displace the fluorescent Fluormone™ PPARγ Green Ligand from the PPARγ-LBD/Fluormone™ PPARγ Green complex, causing the fluorescent ligand to tumble rapidly during its fluorescence lifetime, resulting in a low polarization value. Noncompetitors will not displace the fluorescent ligand from the complex, so the polarization value remains high. The shift in polarization value in the presence of the test compounds is used to determine relative affinity of test compounds for the PPARγ-LBD.
Specifications
Specifications
| Color | Green |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Microplate Reader |
| Quantity | 800 x 20 μL |
| Packaging | 384-well Plate |
| Content And Storage | PV3356 — Fluormone™ PPARγ Green (-20°C) PV4546 — PPARγ-LBD (-80°C) PV3358 — PPARγ Green Screening Buffer (room temperature) P2325 — 1M DTT (-80°C or -20°C) |
| Assay Entry | Biochemical competitive binding |
| Excitation/Emission | 485/535 |
| Gene ID (Entrez) | 5468 |
| Readout | End Point |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?