Learn More
Motic™ Simple Add-on Analyzer Filter
Click to view available options
For Use With (Equipment):
BA310 Series Microscope
Unit Size:
Each
Description
- For use with BA210/310 microscope
- Requires use with Polarizer
- Produces refracted light while polarizer illuminates the specimen
Order Info
ForUseWithEquipment
Specifications
Specifications
| Type | Analyzer Filter |
| Description | Requires use with Polarizer Filter |
| For Use With (Application) | Restricting detected light |
| For Use With (Equipment) | BA310 Series Microscope |
| Quantity | 1 Ea. |