missing translation for 'onlineSavingsMsg'
Learn More

Polaprezinc, MedChemExpress

Product Code. 16494457
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494457 50 mg 50mg
16478588 10 mg 10mg
16428436 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494457 Supplier Medchem Express Supplier No. HYB0729
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Polaprezinc is an orally bioavailable chelate composed of zinc and L-carnosine, with potential gastroprotective, anti-oxidant, anti-ulcer and anti-inflammatory activities.

Polaprezinc is an orally bioavailable chelate composed of zinc and L-carnosine, with potential gastroprotective, anti-oxidant, anti-ulcer and anti-inflammatory activities.

Specifications

Chemical Name or Material Polaprezinc
Physical Form Solid
CAS 107667-60-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C9H11N4O3Zn
For Use With (Application) COVID-19-immunoregulation
Synonym Zinc L-carnosine
Solubility Information H2O : 3.37 mg/mL (11.68 mM; ultrasonic and warming and adjust pH to 1-2 with HCl) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C1[C@@H]([N-](C2=O)[Zn+2](NCC2)[O-]1)CC3=CN=CN3
Molecular Weight (g/mol) 288.59
Formula Weight 288.59
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.