missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLA1, Mouse anti-Human, Clone: 3C11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005422-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: QFSRTGPLCPACMKATLQPEYSDKSLYTQLCFYRYIFDAECALEKLTTDHEKDKLKKQFFTPKVLQDYRKLKNTAEQFLSRSGYSEVNLSKLFAGCAVKSSpecifications
| POLA1 | |
| Monoclonal | |
| Unconjugated | |
| polymerase (DNA directed), alpha 1, catalytic subunit | |
| DKFZp686K1672/POLA/p180 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3C11 | |
| In 1x PBS, pH 7.4 | |
| NM_016937 | |
| POLA1 | |
| POLA (NP_058633, 1363 a.a. ∼ 1462 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5422 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction