missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pokemon Recombinant Rabbit Monoclonal Antibody (JE34-74)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA556485

Product Code. 30275548

  • £434.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pokemon Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

Plays a key role in the instruction of early lymphoid progenitors to develop into B lineage by repressing T-cell instructive Notch signals. Specifically represses the transcription of the CDKN2A gene. Efficiently abrogates E2F1-dependent CDKN2A transactivation/de-repression. Binds to the consensus sequence 5'-[GA][CA]GACCCCCCCCC-3'.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pokemon
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
O95365
ZBTB7A
Synthetic peptide within Human ZBTB7A aa 500 to the C-terminus.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
JE34-74
Unconjugated
ZBTB7A
9030619K07Rik; 9130006G12Rik; AI452336; cLRF; DKFZp547O146; factor binding IST protein 1; Factor that binds to inducer of short transcripts protein 1; FBI1; FBI-1; HIV-1 1st-binding protein 1; HIV-1 inducer of short transcripts binding protein; leukemia/lymohoma related factor cLRF; leukemia/lymphoma related factor; leukemia/lymphoma related factor cLRF; leukemia/lymphoma-related factor; LRF; lymphoma related factor; MGC99631; Oczf; Osteoclast-derived zinc finger protein; POK erythroid myeloid ontogenic factor; Pokemon; POZ and Krueppel erythroid myeloid ontogenic factor; TIP21; TTF-I-interacting peptide 21; Zbtb7; ZBTB7A; zi; zinc finger and BTB domain containing 7a; zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcripts binding protein; zinc finger and BTB domain-containing protein 7A; Zinc finger protein 857A; ZNF857A
Rabbit
Protein A
RUO
51341
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.