missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pokemon Recombinant Rabbit Monoclonal Antibody (3T2J1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537998
This item is not returnable.
View return policy
Description
Positive Samples: HT-29, MCF7 Immunogen sequence: NYDLKNHMRV HTGLRPYQCD SCCKTFVRSD HLHRHLKKDG CNGVPSRRGR KPRVRGGAPD PSPGATATPG APAQPSSPDA RRNGQEKHFK DEDEDEDVAS P.
Plays a key role in the instruction of early lymphoid progenitors to develop into B lineage by repressing T-cell instructive Notch signals. Specifically represses the transcription of the CDKN2A gene. Efficiently abrogates E2F1-dependent CDKN2A transactivation/de-repression. Binds to the consensus sequence 5'-[GA][CA]GACCCCCCCCC-3'.
Specifications
| Pokemon | |
| Recombinant Monoclonal | |
| 0.15 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95365 | |
| ZBTB7A | |
| A synthetic peptide corresponding to a sequence within amino acids 450-550 of human ZBTB7A/FBI-1/LRF (O95365). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3T2J1 | |
| Unconjugated | |
| ZBTB7A | |
| 9030619K07Rik; 9130006G12Rik; AI452336; cLRF; DKFZp547O146; factor binding IST protein 1; Factor that binds to inducer of short transcripts protein 1; FBI1; FBI-1; HIV-1 1st-binding protein 1; HIV-1 inducer of short transcripts binding protein; leukemia/lymohoma related factor cLRF; leukemia/lymphoma related factor; leukemia/lymphoma related factor cLRF; leukemia/lymphoma-related factor; LRF; lymphoma related factor; MGC99631; Oczf; Osteoclast-derived zinc finger protein; POK erythroid myeloid ontogenic factor; Pokemon; POZ and Krueppel erythroid myeloid ontogenic factor; TIP21; TTF-I-interacting peptide 21; Zbtb7; ZBTB7A; zi; zinc finger and BTB domain containing 7a; zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcripts binding protein; zinc finger and BTB domain-containing protein 7A; Zinc finger protein 857A; ZNF857A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51341 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction