missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POGZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552609
This item is not returnable.
View return policy
Description
Immunogen sequence: PEELLTPLAP ALPSPASTAT PPPTPTHPQA LALPPLATEG AECLNVDDQD EGSPVTQEPE LASGGGGSGG VGKKEQLSVK KLRVVLFALC CNTEQAAEHF RNPQRRIRRW LRRFQASQGE NLEGKYLSFE AEEKLAEWV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
POGZ appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid systemThe protein encoded by this gene appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid system. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| POGZ | |
| Polyclonal | |
| Unconjugated | |
| POGZ | |
| 9530006B08Rik; KIAA0461; MRD37; Nbla00003; pogo transposable element derived with ZNF domain; pogo transposable element with ZNF domain; POGZ; putative protein product of Nbla00003; SUHW5; Suppressor of hairy wing homolog 5; WHSUS; Zinc finger protein 280E; zinc finger protein 635; ZNF280E; ZNF635; ZNF635m | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23126 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z3K3 | |
| POGZ | |
| Recombinant protein corresponding to Human POGZ. Recombinant protein control fragment (Product #RP-89484). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction