missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POGZ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-85545
This item is not returnable.
View return policy
Description
POGZ Polyclonal specifically detects POGZ in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| POGZ | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| KIAA0461ZNF635, pogo transposable element with ZNF domain, putative protein product of Nbla00003, SUHW5MGC71543, Suppressor of hairy wing homolog 5, Zinc finger protein 280E, Zinc finger protein 635, ZNF280EZNF635m | |
| Rabbit | |
| 154.91 kDa | |
| 0.1 mg | |
| Primary | |
| This antibody is known to react with mouse POGZ protein. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1.6 mg/mL | |
| Western Blot 1:2000-1:20000, ELISA 1:100000-1:400000 | |
| Q8BZH4 | |
| POGZ | |
| This affinity purified POGZ Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the N-terminus of mouse POGZ protein. (Uniprot: Q8BZH4) | |
| Affinity Purified | |
| RUO | |
| 23126 | |
| Mouse, Rat, Bovine, Canine, Chicken, Opossum, Monkey, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction