missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POGLUT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558140
This item is not returnable.
View return policy
Description
Immunogen sequence: VQELLQFVKA NDDVAQEIAE RGSQFIRNHL QMDDITCYWE NLLSEYSKFL SYNVTRRKGY DQIIPKMLKT EL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
KTELC1 also termed as CLP46 is a glucosyltransferase, which implicates in Protein modification and glycosylation. KTELC1 encodes a 392 amino acids protein and its molecular weight is 46 kDa, but is exists glycosylation modification. KTELC1 is localized in endoplasmic reticulum lumen and abundantly expressed in liver, brain, heart, skeletal muscle, spleen, kidney, placenta, lung and peripheral blood leukocyte. KTELC1 may regulation in cell proliferation and Notch signal pathway.
Specifications
| POGLUT1 | |
| Polyclonal | |
| Unconjugated | |
| POGLUT1 | |
| 9630046K23Rik; C3orf9; CAP10-like 46 kDa protein; CAP10-like protein; CLP46; hCLP46; hRumi; KDELC family like 1; KDELCL1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; KTELC1; MDS010; MDSRP; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Poglut1; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; RGD1306248; Rumi; UNQ490/PRO1006; Wing-shaped neural plate protein; Wsnp; x 010 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NBL1 | |
| POGLUT1 | |
| Recombinant protein corresponding to Human POGLUT1. Recombinant protein control fragment (Product #RP-96395). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction